Infrared Thermometer, Newark/Element
Supplier: Newark
FLUKE-62 MAX
76250-086EA 447.58 USD
76250-086
Infrared Thermometer, Newark/Element
Thermometers Digital Thermometers
Small and lightweight; clips to your tool belt or belt loop or easily fits in to tool box. Distance to spot: Precise laser technology makes for more accurate and repeatable measurements.
  • Dust and water-resistant
  • Small and lightweight
  • Hi and Lo alarms for rapid display of measurements outside the limits
  • Powered by a single, standard AA battery
  • Large screen makes data easier to read, even in dark areas
  • Ergonomically designed completely redesigned for a more natural hand fit
Min/max/avg/dif: Displays the minimum, maximum or average temperature, or the difference between two measurements.
Certifications: IP54 rated for dust and water resistance.