Tamper Seal Labels, Brady
Supplier: Brady
13800
76241-662PK 98.78 USD
76241-662
Tamper Seal Labels, Brady
Labels
Tamper seals are designed for sealing access points, protecting adjustments and sealing enclosures.
  • Protect your warranty exposure by sealing access points with tamper-indicating Tamper Seals
  • High performance white matte vinyl with permanent adhesive is ideal for applications that require resistance to product tampering
Use caution when removing from liner as material is fragile.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
Caution: Labels should not be for consumer goods or consumer warranties per the Federal Trade Commission's interpretation of the Magnuson-Moss Warranty Act. For consumer applications, Brady recommends TL-PK-3 and TL-PK-13.