Solder Tape 5414, 3M
Supplier: 3M
7010334071 7010311734 7010334071
76242-400CS 699.06 USD
76242-400 76242-466 76242-468
Solder Tape 5414, 3M
Tapes
3M™ Water soluble synthetic adhesive wave solder tape 5414 has poly-vinyl alcohol backing which is water soluble and a synthetic water soluble adhesive. This tape is designed to mask gold fingers on printed circuit boards during wave soldering.
  • No adhesive residue after removal
  • Low static upon liner release and when taping board
Humidity extremes do not affect tape properties, used to mask gold fingers on printed circuit boards during wave soldering.
Backing material: Polyvinyl alcohol film