Shoe Cover, Valutek
Supplier: Valutek
VTSHCVAS-LG
76240-976CS 77 USD
76240-976
Shoe Cover, Valutek
Disposable Garments
Disposable anti-skid SPP shoe cover is manufactured with 100% non-linting, non-woven spunbond polypropylene fabric with stitched seams and sprayed latex anti-skid treads on the bottom for safety.
  • Critical environment compatible
  • Part number and lot number traceable
The shoe covers are lightweight, strong and durable. Are tested and manufactured in ISO compliant facilities under Valutek inspection and stringent process control to ensure quality standard and product specifications compliance.
Recommended for use in a cleanroom Class 10000+ (ISO 7+) critical environment. It is also commonly used for protecting against dirt, grime and certain dry particulates in nonhazardous environments.