Shim Stock Sheets, Grainger
Supplier: Grainger
5GE92 5GE94
76246-360EA 159.76 USD
76246-360 76246-368
Shim Stock Sheets, Grainger
Index Cards
For light-duty alignments, it is easy to cut, oil-resistant, and nonconductive to electricity. Minimal compression during use. Made up of PVC.
Applications include use in Conventional Power Plants, Nuclear Plants, Oil and Gas Refineries and Desalinization Projects where corrosion resistance and hardness are required.