Tape 9731, Double Coated, 3M
Supplier: 3M
7010374023
76245-814CS 17300.36 USD
76245-814
Tape 9731, Double Coated, 3M
Tapes
The tape is developed with a high-performance acrylic adhesive on the face side and a silicone adhesive on the liner side of a 1 mil clear PET carrier.
  • Double linered – Adhesive 350
  • Ideal for attaching silicone rubber elastomers and foam
Features acrylic and silicone adhesives for specialized applications such as silicone gasketing. A thin polyester carrier provides dimensional stability and improved handling with ease of die cutting and lamination compared to adhesive transfer tapes.
It is excellent for demanding applications where silicone materials must be bonded to a range of substrates.