Mitutoyo Feeler Gauges, Global Industrial
950-252
76248-660EA 87.48 USD
76248-660
Mitutoyo Feeler Gauges, Global Industrial
This Mitutoyo thickness gauge has tapered blades with 1/4" width at tip. Each leaf is marked with thickness and each leaf can be detached.
  • Tapered leaves to get into tight places
  • Knurled lock screw
  • Protective outer case with thumb slot
  • Leaves are detachable from set
  • 3" blade length