Fiber Optic Illuminator, Scienscope International
Supplier: Scienscope International
IL-FOI-150
76238-854EA 869.82 USD
76238-854
Fiber Optic Illuminator, Scienscope International
This features "cold" illumination through filtration and a series of user defined colored filters. The cold light source has high intensity fiber optic illumination and accessories that allow you to position your light for best possible viewing.
  • New voltage reader
  • New design for easy lamp replacement
  • Protection against overload
  • Ideal for microscope applications
  • Internal light filters holder
The original design shown above features a sturdy small footprint through integrated solid state circuitry. The lamp is equipped with a variable control. This provides the best illumination for high brightness and low temperatures.
Brightness of 23000 LU and color temperature 3200 k (at full output).