HALYARD Protective Surgical Caps, NDC
69520
76241-122CS 130.3 USD
76241-122
HALYARD Protective Surgical Caps, NDC
This cap is made of a tri-layer fabric on the sides and spunbond fabric on the top.
For use when there is a risk of light fluid contamination from splashing or spraying.