Miniature and Micro Miniature Brushes, Brass, Tanis Inc
Supplier: Tanis
04986 04994 4988 05190
76254-334PK 109.59 USD
76254-334 76254-268 76254-336 76254-340
Miniature and Micro Miniature Brushes, Brass, Tanis Inc
Brushes
Miniature brushes with loop for hand use. Single stem and spiral loop end.
  • Stem Ø 0.62"
  • Filament Ø 0.003"
Used for ultra-fine cross hole deburring, cleaning, blending and polishing of small diameter holes.