Repositionable Vinyl Cloth Inspection Arrows, Brady
Supplier: Brady
49346 49343
76244-624EA 5.95 USD
76244-624 76245-856
Repositionable Vinyl Cloth Inspection Arrows, Brady
Labels
Pin-point rework areas on printed circuit boards with these durable yet repositionable vinyl cloth inspection arrows.
  • Small, brightly colored arrows draw the eye to the specific point of need
  • Excellent flexibility for wrapping around curved surfaces
  • Withstands moderate heat, oil, water and dirt
  • Withstands –40 to 176 °F
Repositionable vinyl cloth holds tightly to surfaces but allows item to be repositioned with ease if necessary.
Abrasion, corrosion, humidity, UV and weather resistant.