Double-Acting Air Cylinders, Zoro

Supplier: Zoro
5TRE3
76249-780EA 192.45 USD
76249-780
Double-Acting Air Cylinders, Zoro
Gas Cylinders
This is 9/16" bore round double acting air cylinder with 3" stroke.

  • Stainless steel body and piston
  • Silver finish
  • Round non-repairable
  • Delrin end cap

Temp. Range: 32 to 160 °F.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR