Would you like to log in again?
VWR® Sealing Film Roll with Pre-Scored Sheets
Supplier: VWR International
VWR®
SP2-RLNL-500
76293-222RL 221.59 USD InStock
76293-222
VWR® Sealing Film Roll with Pre-Scored Sheets
Seals Microplate Sealing Films and Foils
An economical, efficient way to rapidly seal microplates; faster than applying single sheeted film.
  • Ideal for ELISA, incubation, and general use
  • 2 mil polyester film minimizes evaporation, reduces cross-contamination, and prevents spillage
  • Acrylic adhesive
  • A self-standing, non-slip dispenser is supplied for ease of use
It is useful as a simple cover to protect samples from external contamination, dust, and debris.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty