3,3'-Diaminobenzidine tetrahydrochloride (DAB), EM Grade

Supplier: Electron Microscopy Sciences
Warning

Synonyms: DAB HCl3,3',4,4'-Biphenyltetramine tetrahydrochloride

13080
100503-804EA 42.94 USD
100503-804
3,3'-Diaminobenzidine tetrahydrochloride (DAB), EM Grade
3,3'-Diaminobenzidine tetrahydrochloride

Used for histochemical localization of peroxidase with DAB and in immunocytochemistry (Sternberger, et.al., J. Histochem, Cytochem., 18,215 (1970)).


  • Avaialble also in pre-weighed (10 mg DAB 4HCl) and packed in tablet form, to facilitate your work.

Formula: C₁₂H₁₈Cl₄N₄
MW: 360.11 g/mol
Melting Pt: 300 °C (572 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00012969
CAS Number: 7411-49-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR