Allevi 2 3D Bioprinter, Two Extruder System, Allevi
Supplier: Allevi
A2
76292-706EA 22896 USD
76292-706
Allevi 2 3D Bioprinter, Two Extruder System, Allevi
Printers Bioprinters
The Allevi 2 is the world's first desktop 3D bioprinter that prints living tissue out of human cells. It is a beautifully designed, precision manufactured 3D bioprinter that prints cells and bioinks. The Allevi 2 is the most versatile bioprinter on the market, trusted by leading researchers worldwide, and is easy enough for everyone in the lab to use. The Allevi 2 is ready to turn your biggest ideas into your greatest work.
  • Two extruder system
  • Small footprint fits under any sterile hood
  • Accessibility - print both soft and hard tissues
  • Precision engineered - 10 micron on each axis
  • Easy to use
  • Visible Light Photocuring
  • Temperature control from RT-160C
Allevi 2 is the bioprinter for researchers, by researchers. It is the most versatile bioprinter on the market, trusted by leading researchers worldwide, and is easy enough for everyone in the lab to use.
Designed with a small footprint that fits under any sterile hood, the Allevi 2 is able to print living cells under sterile conditions. A temperature range from RT-160C allows you to bioprint a wide range of biomaterials including cells, hydrogels, and thermoplastics. The open system accepts standard 10ml luer lock syringes so you have complete control over your bioprints.
Users in over 200 labs use this bioprinter to publish game-changing work in the fields of tissue engineering, material science, regenerative medicine, pharmacology and biomedical devices. Allevi 2 is built on groundbreaking ideas.
Certifications: ISO-9001 Certified.
Ordering information: Allevi Bioink Starter Kit, basic software and air compressor included.