Cynomolgus Recombinant IFNA13/Interferon alpha-13 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90108-C08H-100 90108-C08H-20 90108-C08H-5 90108-C08H-20 90108-C08H-5
103585-326EA 351.48 USD
CA76453-994 103585-326 103678-050 103670-330 CA76453-182
Cynomolgus Recombinant IFNA13/Interferon alpha-13 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus IFNA13 (G7NFW4) (Met1-Glu190) was expressed with a polyhistidine tag at the C-terminus.