Mouse Recombinant SELPLG (from HEK293 cells)
Supplier: Sino Biological
50770-M02H-50
103668-094EA 178 USD
103668-094
Mouse Recombinant SELPLG (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of mouse SELPLG (Q62170) (Met 1-Cys 307) was fused with the Fc region of human IgG1 at the C-terminus.