MERS-CoV Recombinant Spike/RBD Protein fragment (RBD, aa 367-606, rFc Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40071-V31B1-100
103632-832EA 452.48 USD
103632-832
MERS-CoV Recombinant Spike/RBD Protein fragment (RBD, aa 367-606, rFc Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the receptor binding domain (RBD) of MERS-CoV spike protein fragment (AFS88936.1) (Glu367-Tyr606) was fused with the Fc region of rabbit IgG at the C-terminus.