Human Recombinant EIF-5A/EIF5 Protein (GST Tag) (from E. coli)
Supplier: Sino Biological
13932-H09E-100 13932-H09E-20
103614-324EA 116.1 USD
103614-324 103623-536
Human Recombinant EIF-5A/EIF5 Protein (GST Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human EIF5 (P55010) (Met1-Asp150) was fused with the GST tag at the N-terminus.