Human Recombinant TFRC (from HEK293 Cells), His Tag
Supplier: Sino Biological
11020-H07H-100 11020-H07H-50 11020-H07H-1 11020-H07H-50
103680-380EA 178 USD
103680-380 103623-722 CA76455-912 76642-154
Human Recombinant TFRC (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human TFRC (NP_003225.2) (Cys 89-Phe 760) was expressed with a polyhistidine tag at the N-terminus.