Human Recombinant MESDC2/MESD Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10949-H08H-50 10949-H08H-50 10949-H08H-100
103621-690EA 300.98 USD
CA76456-742 103621-690 103681-292
Human Recombinant MESDC2/MESD Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human MESDC2 (NP_055969.1) (Ala 34-Lys 230) was fused with a signal peptide at the N-terminus and a polyhistidine tag at the C-terminus.