Human Recombinant IL10Rb Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10945-H08H-20 10945-H08H-100 10945-H08H-20 10945-H08H-1
103680-836EA 148.35 USD
103680-836 CA76456-306 76641-370 103621-680
Human Recombinant IL10Rb Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domainof human IL10Rβ (NP_000619.3) (Met 1-Ser 220) was expressed, fused with a polyhistidine tag at the C-terminus.