Human Recombinant GDF15 (from HEK293 Cells)
Supplier: Sino Biological
10936-H01H-20
CA76455-800
Human Recombinant GDF15 (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of human GDF15 (NP_004855.2) (Ala 197-Ile 308) was expressed with the fused Fc region of human IgG1 at the N-terminus.