Human Recombinant TIMP-1/TIMP1 Protein (from HEK293 Cells)
Supplier: Sino Biological
10934-HNAH-20 10934-HNAH-10 10934-HNAH-20 10934-HNAH-10
103680-264EA 148.35 USD
103680-264 CA76455-796 103680-266 CA76455-798
Human Recombinant TIMP-1/TIMP1 Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of human TIMP1 (NP_003245.1) (Cys 24-Ala 207) was expressed.