Human Recombinant TIMP1 (from HEK293 Cells), His Tag
Supplier: Sino Biological
10934-H08H-10
CA76455-792
Human Recombinant TIMP1 (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human TIMP1 (NP_003245.1) (Met 1-Ala 207) with a C-terminal polyhistidine tag was expressed.