Human Recombinant TIGIT (from HEK293 Cells)
Supplier: Sino Biological
10917-H02H-200
103621-642EA 448 USD
103621-642
Human Recombinant TIGIT (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human TIGIT (NP_776160.2) (Met1-Pro141) was expressed with the Fc region of human IgG1 at the C-terminus.