Human Recombinant XCL1 (from Baculovirus (Insect Cells)), His Tag
Supplier: Sino Biological
10672-H08B-20
CA76455-322
Human Recombinant XCL1 (from Baculovirus (Insect Cells)), His Tag
Proteins and Peptides
A DNA sequence encoding the human XCL1 (P47992) (Met 1-Gly 114) was expressed, fused with a polyhistidine tag at the C-terminus.