Human Recombinant CAMK4/CaMKIV Protein (GST Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
10664-H09B-50 10664-H09B-20 10664-H09B-20 10664-H09B-50
103679-656EA 199.98 USD
CA76455-186 CA76455-188 103679-656 103679-654
Human Recombinant CAMK4/CaMKIV Protein (GST Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human CAMK4 (NP_001735.1) (Met 1-Tyr 473) was fused with the GST tag at the N-terminus.