Human Recombinant CCL23 (from Yeast), His Tag
Supplier: Sino Biological
10655-H08Y-5
CA76453-604
Human Recombinant CCL23 (from Yeast), His Tag
Proteins and Peptides
A DNA sequence encoding the human CCL23 (P55773.3) (Arg46-Asn120) was expressed with a polyhistidine tag at the C-terminus.