Human Recombinant CCL28 (from E. coli), His Tag
Supplier: Sino Biological
10618-H07E-20
CA76455-134
Human Recombinant CCL28 (from E. coli), His Tag
Proteins and Peptides
A DNA sequence encoding the human CCL28 (NP_683513.1) (Ile23-Tyr127) was expressed with a polyhistidine tag at the N-terminus.