Human Recombinant IL36A (from E. coli)
Supplier: Sino Biological
10607-HNCE1-10
103622-534EA 191.35 USD
103622-534
Human Recombinant IL36A (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human IL1F6 (Q9UHA7) (Ala6-Phe158) was expressed and purified, with two additional amino acids (Gly & Pro) at the N-terminus.