Human Recombinant ACVR1B (from HEK293 cells)
Supplier: Sino Biological
10583-H03H-100 10583-H03H-200
103622-516EA 191.35 USD
103622-516 103622-518
Human Recombinant ACVR1B (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the human ACVR1B (NP_004293.1) extracellular domain (Met 1-Glu 126) was fused with C-terminal His-tagged Fc region of human IgG1 at the C-terminus.