Human Recombinant ANGPTL4 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10563-H01H-100 10563-H01H-50 10563-H01H-50
103622-498EA 300.98 USD
103622-498 103679-982 CA76455-514
Human Recombinant ANGPTL4 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human ANGPTL4 (Q9BY76) (Pro166-Ser406) was expressed,with the fused Fc region of human IgG1 at the N-terminus.