Human Recombinant DC-SIGNR/CD299/CLEC4M Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10559-H01H-50 10559-H01H-50 10559-H01H-100
103622-492EA 351.48 USD
103622-492 CA76454-514 103678-982
Human Recombinant DC-SIGNR/CD299/CLEC4M Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Ser 78-Glu 399) of human DC-SIGNR (NP_055072.3) was fused with the Fc region of human IgG1 at the N-terminus.