Human Recombinant PDGFRA (from HEK293 Cells)
Supplier: Sino Biological
10556-HCCH-50 10556-HCCH-50
103678-936EA 178 USD
103678-936 CA76454-482
Human Recombinant PDGFRA (from HEK293 Cells)
Proteins and Peptides
The mature form of human PDGFRa (NP_006197.1) extracellular domain (Met 1-Glu 524) with five amino acids (DDDDK) at the C-terminus was expressed and purified.