Human PDGFRa / CD140a Protein (His & GST Tag)
Supplier: Sino Biological
10556-H20B1-50
103622-488EA 277.35 USD
103622-488
Human PDGFRa / CD140a Protein (His & GST Tag)
Proteins and Peptides
A DNA sequence encoding the human PDGFRA (P16234-1) (Gln551-Leu1089) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.