Human Recombinant BMPR2/BMPR-II Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10551-H08H-50 10551-H08H-100 10551-H08H-1 10551-H08H-50
103679-520EA 148.35 USD
103679-520 76640-928 103621-310 CA76455-052
Human Recombinant BMPR2/BMPR-II Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human BMPR-II (NP_001195.2) extracellular domain (Met 1-Ile 151) was expressed with a C-terminal polyhistidine tag.