Human Recombinant ACY1 (from Baculovirus (Insect cells)), His Tag
Supplier: Sino Biological
10549-H08B-20
CA76455-044
Human Recombinant ACY1 (from Baculovirus (Insect cells)), His Tag
Proteins and Peptides
A DNA sequence encoding the full length of human ACY1 (NP_000657.1) (Met 1-Ser 408) was expressed with a polyhistidine tag at the C-terminus.