Human Recombinant VEGFC (from HEK293 Cells), His Tag
Supplier: Sino Biological
10542-H08H-100 10542-H08H-50 10542-H08H-1 10542-H08H-50 10542-H08H-5
103621-302EA 300.98 USD
103621-302 103679-504 76641-790 CA76455-036 103678-954
Human Recombinant VEGFC (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the mature form of human VEGFC (NP_005420.1) corresponding to amino acid (Thr 103-Arg 227) was expressed with a C-terminal polyhistidine tag.