Human Recombinant FGFR4 (from HEK293 cells)
Supplier: Sino Biological
10538-H02H-50
CA76454-480
Human Recombinant FGFR4 (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 (NP_002002.3) precursor was fused with the Fc region of human IgG1 at the C-terminus.