Human Recombinant IL17RD Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10507-H08H-50 10507-H08H-100 10507-H08H-50
103679-462EA 191.35 USD
103679-462 CA76454-994 103621-288
Human Recombinant IL17RD Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human IL17RD (NP_060033.3) precursor (Met 1-Arg 299) was expressed with a C-terminal polyhistidine tag.