Human Recombinant TNFRSF4/OX40/CD134 Protein (His Tag) , Biotinylated (from HEK293 Cells)
10481-H08H-B-20
103632-618EA 187.88 USD
103632-618
Human Recombinant TNFRSF4/OX40/CD134 Protein (His Tag) , Biotinylated (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met1-Ala216) of human TNFRSF4 (NP_003318.1) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.