Human Recombinant TNFRSF4 (from HEK293 Cells), His Tag
Supplier: Sino Biological
10481-H08H-50 10481-H08H-100 10481-H08H-50 10481-H08H-1
103679-396EA 178 USD
CA76454-928 103679-396 76641-748 103621-278
Human Recombinant TNFRSF4 (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Ala 216) of human TNFRSF4 (NP_003318.1) was expressed, fused with a polyhistidine tag at the C-terminus.