Human Recombinant Frizzled-5/FZD5 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10473-H08H-50 10473-H08H-100 10473-H08H-50 10473-H08H-1
76641-618EA 2268 USD
76641-618 CA76454-896 103679-364 103621-274
Human Recombinant Frizzled-5/FZD5 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Pro 167) of human frizzled 5 (NP_003459.2) was fused with a polyhistidine tag at the C-terminus.