Human Recombinant TRAIL R2/CD262/TNFRSF10B Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10465-H08H-1 10465-H08H-200 10465-H08H-100
103621-258EA 250.48 USD
103621-258 103621-260 76641-002
Human Recombinant TRAIL R2/CD262/TNFRSF10B Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human TNFRSF10B (NP_003833.4) extracellular domain (Met1-Glu182) was expressed with a polyhistidine tag at the C-terminus.