Human Recombinant FAP (from HEK293 cells), Biotin, His Tag
Supplier: Sino Biological
10464-H07H-B-10
103632-616EA 298 USD
103632-616
Human Recombinant FAP (from HEK293 cells), Biotin, His Tag
Proteins and Peptides
A DNA sequence encoding the human FAP isoform 1 (Q12884-1) extracellular domain (Leu26-Asp760) was fused with the polyhistidie-tag at the N-terminus. The purified protein was biotinylated in vitro.