Human Recombinant Carbonic Anhydrase XIII/CA13 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
10461-H08E-50 10461-H08E-100 10461-H08E-50
103621-248EA 250.48 USD
103621-248 103679-352 CA76454-884
Human Recombinant Carbonic Anhydrase XIII/CA13 Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human CA13 (NP_940986.1) (Met 1-His 262) was expressed, with a polyhistide tag at the C-terminus.