Human Recombinant BMPR1B/ALK-6 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10460-H02H-100 10460-H02H-200
103621-246EA 452.48 USD
103621-246 103621-244
Human Recombinant BMPR1B/ALK-6 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human ALK6 (NP_001194.1)(Lys14-Arg126) was expressed with the Fc region of human IgG1 at the C-terminus.