Human Recombinant CA14 (from HEK293 cells), His Tag
Supplier: Sino Biological
10458-H08H-10 10458-H08H-10 10458-H08H-20 10458-H08H-20
103679-338EA 266.6 USD
CA76454-870 CA76454-868 103679-338 103679-336
Human Recombinant CA14 (from HEK293 cells), His Tag
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human CA14 (NP_036245.1) (Met 1-Met 290) was expressed with a C-terminal polyhistidine tag.