Human Recombinant BMPRIA/ALK-3/CD292 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10446-H08H-200 10446-H08H-100
103621-224EA 374.1 USD
103621-224 103621-222
Human Recombinant BMPRIA/ALK-3/CD292 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Arg 152) of human ALK3 (NP_004320.2) was fused with a polyhistidine tag at the C-terminus.